Anti-alpha II Spectrin Mouse Monoclonal Antibody [clone: 3D7]

Supplier: Antibodies.com
A85353-100 A85353-50
77080-826EA 877.69 CAD
77080-826 77069-234
Anti-alpha II Spectrin Mouse Monoclonal Antibody [clone: 3D7]
Antibodies
Mouse monoclonal (3D7) antibody to alpha II Spectrin for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:3000, ICC/IF: 1:500, IHC: 1:50

Type: Primary
Antigen: alpha II Spectrin
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR