Anti-mCherry Chicken Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85304-100 A85304-50
77078-454EA 877.69 CAD
77078-454 77068-802
Anti-mCherry Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to mCherry for WB, ICC/IF and IHC.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:2000 to 1:5000, ICC/IF: 1:1000, IHC: 1:1000
  • Host: Chicken
  • Clonality: Polyclonal
  • Isotype: IgY
  • Conjugate: Unconjugat

Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR