Anti-Nestin Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85294-100 A85294-50
77067-634EA 877.69 CAD
77067-634 77068-186
Anti-Nestin Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Nestin for WB and ICC/IF with samples derived from Human.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:2000 to 1:1:500

Type: Primary
Antigen: Nestin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR