Anti-Adenylate Cyclase 3 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85287-100 A85287-50
77061-834EA 877.69 CAD
77061-834 77074-820
Anti-Adenylate Cyclase 3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Adenylate Cyclase 3 for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1000, ICC/IF: 1:10000 to 1:20000, IHC: 1:10000 to 1:2000

Type: Primary
Antigen: Adenylate Cyclase 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR