Anti-MMP7 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84120-100
77071-584EA 797.27 CAD
77071-584
Anti-MMP7 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MMP7 for ELISA, WB and IHC with samples derived from Human.

  • Validated Applications: ELISA, WB, IH

Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR