Anti-IgG Antibody

Supplier: ANTIBODIES.COM LLC
A190-200 A190-100
77041-690EA 1022.73 CAD
77041-690 77041-688
Anti-IgG Antibody
Antibodies
  • Validated Applications: ELISA, WB
  • Recommended Dilutions: ELISA: 1/1000 to 1/5000, WB: 1/500 to 1/10000

Type: Primary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR