Anti-AFP Mouse Monoclonal Antibody [clone: KT137]

Supplier: ANTIBODIES.COM LLC
A147-200 A147-100
77041-556EA 1022.73 CAD
77041-556 77041-554
Anti-AFP Mouse Monoclonal Antibody [clone: KT137]
Antibodies
Mouse monoclonal (KT137) antibody to AFP for sELISA with samples derived from Human.

  • Validated Applications: sELISA
  • Recommended Dilutions: sELISA: 3 µg/ml

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: KT137
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR