Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: KT112]

Supplier: ANTIBODIES.COM LLC
A138-100 A138-200
77041-522EA 797.27 CAD
77041-522 77041-524
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: KT112]
Antibodies
Mouse monoclonal (KT112) antibody to Ig lambda light chain for ELISA and WB with samples derived from Rat.

  • Validated Applications: ELISA, WB
  • Recommended Dilutions: ELISA: 1/1000, WB: 1/500 to 1/5000

Type: Primary
Antigen: Lambda Light Chain
Clonality: Monoclonal
Clone: KT112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR