Anti-G-CSF Mouse Monoclonal Antibody [clone: KT27]

Supplier: ANTIBODIES.COM LLC
A79-100 A79-200
77041-904EA 797.27 CAD
77041-904 77041-906
Anti-G-CSF Mouse Monoclonal Antibody [clone: KT27]
Antibodies
Mouse monoclonal (KT27) antibody to G-CSF for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml, WB: 1/500 to 1/10000

Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: KT27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR