Anti-FGF2 Mouse Monoclonal Antibody [clone: KT49]

Supplier: ANTIBODIES.COM LLC
A69-200 A69-100
77041-868EA 1022.73 CAD
77041-868 77041-866
Anti-FGF2 Mouse Monoclonal Antibody [clone: KT49]
Antibodies
Mouse monoclonal (KT49) antibody to FGF2 for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: FGF2
Clonality: Monoclonal
Clone: KT49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR