Anti-IgG Mouse Monoclonal Antibody [clone: KT47]

Supplier: Antibodies.com
A66-200 A66-100
77041-858EA 1022.73 CAD
77041-858 77041-856
Anti-IgG Mouse Monoclonal Antibody [clone: KT47]
Antibodies
Mouse monoclonal (KT47) antibody to IgG for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone: KT47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR