Anti-Endostatin Mouse Monoclonal Antibody [clone: KT58]

Supplier: ANTIBODIES.COM LLC
A64-100 A64-200
77041-848EA 797.27 CAD
77041-848 77041-850
Anti-Endostatin Mouse Monoclonal Antibody [clone: KT58]
Antibodies
Mouse monoclonal (KT58) antibody to Endostatin for sELISA with samples derived from Human.

  • Validated Applications: sELISA
  • Recommended Dilutions: ELISA: 1 µg/ml, sELISA: 2 to 3 µg/ml

Type: Primary
Antigen: Endostatin
Clonality: Monoclonal
Clone: KT58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR