Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: KT39]

Supplier: ANTIBODIES.COM LLC
A41-200 A41-100
77041-764EA 1022.73 CAD
77041-764 77041-762
Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: KT39]
Antibodies
Mouse monoclonal (KT39) antibody to C-reactive protein for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: C Reactive Protein
Clonality: Monoclonal
Clone: KT39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR