Anti-beta 2 Microglobulin Rabbit Recombinant Antibody [clone: B2M/7013R]

Supplier: Antibodies.com LLC
A278052-100
76956-338EA 854.39 CAD
76956-338
Anti-beta 2 Microglobulin Rabbit Recombinant Antibody [clone: B2M/7013R]
Antibodies
Recombinant rabbit monoclonal [B2M/7013R] antibody to beta 2 Microglobulin for IHC-P with samples derived from Human and Non-Human Primates.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Recombinant
Clone: B2M/7013R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Non-human primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR