Anti-Annexin VI Mouse Monoclonal Antibody [clone: 4G3]

Supplier: ANTIBODIES.COM LLC
A270554-100 A270554-50
77078-040EA 877.69 CAD
77078-040 77072-376
Anti-Annexin VI Mouse Monoclonal Antibody [clone: 4G3]
Antibodies
Mouse monoclonal [4G3] antibody to Annexin VI for WB and ICC/IF with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:1,000 to 1:2,000, ICC/IF: 1:1,000

Type: Primary
Antigen: Annexin VI
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR