Anti-Syndecan-1 Rabbit Recombinant Antibody [clone: DM46] (Biotin)

Supplier: ANTIBODIES.COM LLC
A317418-100 A317418-500
77182-306EA 1602.44 CAD
77182-306 77182-308
Anti-Syndecan-1 Rabbit Recombinant Antibody [clone: DM46] (Biotin)
Antibodies
Recombinant rabbit monoclonal [DM46] antibody to Syndecan-1 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: Syndecan-1
Clonality: Recombinant
Clone: DM46
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR