Anti-CD28 Rabbit Recombinant Antibody [clone: DM63] (Biotin)

Supplier: ANTIBODIES.COM LLC
A317402-100 A317402-500
77182-242EA 1602.44 CAD
77182-242 77182-244
Anti-CD28 Rabbit Recombinant Antibody [clone: DM63] (Biotin)
Antibodies
Recombinant rabbit monoclonal [DM63] antibody to CD28 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: CD28
Clonality: Recombinant
Clone: DM63
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR