Anti-CD137 Rabbit Recombinant Antibody [clone: DM67] (Biotin)

Supplier: ANTIBODIES.COM LLC
A317398-100 A317398-500
77182-226EA 1602.44 CAD
77182-226 77182-228
Anti-CD137 Rabbit Recombinant Antibody [clone: DM67] (Biotin)
Antibodies
Recombinant rabbit monoclonal [DM67] antibody to CD137 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: CD137
Clonality: Recombinant
Clone: DM67
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR