Anti-Mesothelin Rabbit Recombinant Antibody [clone: DM71]

Supplier: ANTIBODIES.COM LLC
A317392-100 A317392-500
77182-202EA 1111.21 CAD
77182-202 77182-204
Anti-Mesothelin Rabbit Recombinant Antibody [clone: DM71]
Antibodies
Recombinant rabbit monoclonal [DM71] antibody to Mesothelin for ELISA and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: Mesothelin
Clonality: Recombinant
Clone: DM71
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR