Anti-Von Willebrand Factor Rabbit Recombinant Antibody [clone: VWF/7979R]

Supplier: Antibodies.com LLC
A317172-100
77181-258EA 1244.96 CAD
77181-258
Anti-Von Willebrand Factor Rabbit Recombinant Antibody [clone: VWF/7979R]
Antibodies
Recombinant rabbit monoclonal [VWF/7979R] antibody to Von Willebrand Factor for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Von Willebrand Factor
Clonality: Recombinant
Clone: VWF/7979R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR