Anti-smooth muscle Myosin heavy chain 11 Rabbit Recombinant Antibody [clone: MYH11/8185R]

Supplier: Antibodies.com LLC
A317044-100
77181-002EA 1244.96 CAD
77181-002
Anti-smooth muscle Myosin heavy chain 11 Rabbit Recombinant Antibody [clone: MYH11/8185R]
Antibodies
Recombinant rabbit monoclonal [MYH11/8185R] antibody to smooth muscle Myosin heavy chain 11 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: smooth muscle Myosin heavy chain 11
Clonality: Recombinant
Clone: MYH11/8185R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR