Anti-Sarcomeric alpha Actinin Mouse Recombinant Antibody [clone: rACTN2/7361]

Supplier: Antibodies.com LLC
A316988-100
77180-890EA 1244.96 CAD
77180-890
Anti-Sarcomeric alpha Actinin Mouse Recombinant Antibody [clone: rACTN2/7361]
Antibodies
Recombinant mouse monoclonal [rACTN2/7361] antibody to Sarcomeric alpha Actinin for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Sarcomeric alpha Actinin
Clonality: Recombinant
Clone: rACTN2/7361
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR