Anti-TGN46 Rabbit Monoclonal Antibody [clone: ARC2197]

Supplier: ANTIBODIES.COM LLC
A307687-100
77226-972EA 781.15 CAD
77226-972 77226-974
Anti-TGN46 Rabbit Monoclonal Antibody [clone: ARC2197]
Antibodies
Rabbit monoclonal [ARC2197] antibody to TGN46 for WB, IHC and ICC/IF with samples derived from Human.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: TGN46
Clonality: Monoclonal
Clone: ARC2197
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR