ESD Cleanroom Soft Sole Boots

Supplier: Transforming Technologies
TX40BEWH05 TX40BEWH06 TX40BENB01 TX40BENB02 TX40BENB03 TX40BENB04 TX40BENB05 TX40BENB06 TX40BEWH01 TX40BEWH02 TX40BEWH03 TX40BEWH04
77008-078PR 132.36 CAD
77008-078 77008-100 77008-108 77008-184 77008-210 77008-118 77008-168 77008-074 77008-136 77008-076 77008-156 77008-186
ESD Cleanroom Soft Sole Boots
Workwear
ESD-safe integrated cleanroom wear designed for mobile users in controlled environments.

  • TX4000 ESD grid fabric; 99% polyester/1% carbon
  • Three-layer ESD rubber soft sole (non marking rubber inside and outside)
  • Hypalon side wall; Reusable/launderable
  • Combination 1" elastic and webbing at top of closure with snap adjustment
  • Replaceable 18" internal ground ribbon attachment included, with 24" long ground ribbon available for boots
  • Internal snaps to electrically connect to coverall and ground ribbon
  • Color coded straps denote size; ISO cleanroom: ISO 6 (Class 1000)

The Transforming Technologies TX4000 ESD cleanroom garment system protects the controlled environment from the number one source of contamination and static found in cleanrooms - people. Electrostatic charge is an invisible contaminant that has the potential to degrade or destroy both product and equipment, as well as attract unwanted particles. Controlling these charges on mobile operators with the TX4000 garment system throughout the controlled environment is essential to a productive manufacturing process.

Certifications: Meets standards required for ANSI/ESD S20.20 and STM9.1, in compliance with IEST-RP-CC052.1.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR