Anti-LYVE1 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A83506-100
77075-650EA 797.27 CAD
77075-650
Anti-LYVE1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to LYVE1 for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, WB

Type: Primary
Antigen: LYVE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR