Anti-5 Lipoxygenase/5-LO Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A11051-100 A11051-50
76868-050EA 707.92 CAD
76868-050 76849-018
Anti-5 Lipoxygenase/5-LO Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to 5 Lipoxygenase / 5-LO (phospho Ser271) for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: 5 Lipoxygenase/5-LO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR