Anti-GSK3 alpha (phospho Tyr279) + GSK3 beta (phospho Tyr216) Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A10992-100
76848-674EA 724.75 CAD
76848-674 76867-206
Anti-GSK3 alpha (phospho Tyr279) + GSK3 beta (phospho Tyr216) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GSK3 alpha (phospho Tyr279) + GSK3 beta (phospho Tyr216) for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: GSK3 alpha (phospho Tyr279) + GSK3 beta (phospho Tyr216)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR