Anti-TNF Alpha Mouse Monoclonal Antibody [clone: KT61]

Supplier: ANTIBODIES.COM LLC
A107-200 A107-100
77041-418EA 1022.73 CAD
77041-418 77041-416
Anti-TNF Alpha Mouse Monoclonal Antibody [clone: KT61]
Antibodies
Mouse monoclonal (KT61) antibody to TNF alpha for sELISA with samples derived from Human.

  • Validated Applications: sELISA
  • Recommended Dilutions: ELISA: 1 µg/ml, sELISA: 05 to 2 µg/ml

Type: Primary
Antigen: TNF Alpha
Clonality: Monoclonal
Clone: KT61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR