Anti-TNF Alpha Mouse Monoclonal Antibody [clone: KT31]

Supplier: ANTIBODIES.COM LLC
A105-100 A105-200
77041-410EA 797.27 CAD
77041-410 77041-412
Anti-TNF Alpha Mouse Monoclonal Antibody [clone: KT31]
Antibodies
Mouse monoclonal (KT31) antibody to TNF alpha for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: TNF Alpha
Clonality: Monoclonal
Clone: KT31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR