Anti-Integrin alpha 4/CD49D Recombinant Recombinant Antibody

Supplier: Antibodies.com LLC
A318888-100
77188-020EA 447.54 CAD
77188-020
Anti-Integrin alpha 4/CD49D Recombinant Recombinant Antibody
Antibodies
Recombinant humanized monoclonal [Natalizumab Biosimilar] antibody to Integrin alpha 4/CD49D for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:10

Type: Primary
Antigen: Integrin alpha 4/CD49D
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG4
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR