Anti-Eph Receptor A4/SEK Chimera Recombinant Monoclonal Antibody [clone: DMC472]

Supplier: ANTIBODIES.COM LLC
A318718-100 A318718-500
77187-510EA 1111.21 CAD
77187-510 77187-512
Anti-Eph Receptor A4/SEK Chimera Recombinant Monoclonal Antibody [clone: DMC472]
Antibodies
Recombinant chimeric monoclonal [DMC472] antibody to Eph Receptor A4/SEK for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: Eph Receptor A4/SEK
Clonality: Recombinant
Clone: DMC472
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR