Anti-GPCR GPR87 Chimera Recombinant Monoclonal Antibody [clone: DMC478]

Supplier: ANTIBODIES.COM LLC
A318712-100 A318712-500
77187-486EA 1111.21 CAD
77187-486 77187-488
Anti-GPCR GPR87 Chimera Recombinant Monoclonal Antibody [clone: DMC478]
Antibodies
Recombinant chimeric monoclonal [DMC478] antibody to GPCR GPR87 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: GPCR GPR87
Clonality: Recombinant
Clone: DMC478
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR