Anti-CD80 Rabbit Recombinant Antibody [clone: DM109]

Supplier: ANTIBODIES.COM LLC
A318598-100 A318598-500
77187-030EA 1111.21 CAD
77187-030 77187-032
Anti-CD80 Rabbit Recombinant Antibody [clone: DM109]
Antibodies
Recombinant rabbit monoclonal [DM109] antibody to CD80 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD80
Clonality: Recombinant
Clone: DM109
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR