Anti-CD7 Rabbit Recombinant Antibody [clone: DM118]

Supplier: ANTIBODIES.COM LLC
A318589-500 A318589-100
77186-996EA 3108.33 CAD
77186-996 77186-994
Anti-CD7 Rabbit Recombinant Antibody [clone: DM118]
Antibodies
Recombinant rabbit monoclonal [DM118] antibody to CD7 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD7
Clonality: Recombinant
Clone: DM118
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR