Anti-BAFF-R Rabbit Recombinant Antibody [clone: DM143]

Supplier: Antibodies.com
A318565-100 A318565-500
77186-898EA 1111.21 CAD
77186-898 77186-900
Anti-BAFF-R Rabbit Recombinant Antibody [clone: DM143]
Antibodies
Recombinant rabbit monoclonal [DM143] antibody to BAFF-R for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: BAFF-R
Clonality: Recombinant
Clone: DM143
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR