Anti-TACI Rabbit Recombinant Antibody [clone: DM151]

Supplier: ANTIBODIES.COM LLC
A318557-100 A318557-500
77186-866EA 1111.21 CAD
77186-866 77186-868
Anti-TACI Rabbit Recombinant Antibody [clone: DM151]
Antibodies
Recombinant rabbit monoclonal [DM151] antibody to TACI for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: TACI
Clonality: Recombinant
Clone: DM151
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR