Anti-IL-6R Rabbit Recombinant Antibody [clone: DM152]

Supplier: ANTIBODIES.COM LLC
A318556-100 A318556-500
77186-862EA 1111.21 CAD
77186-862 77186-864
Anti-IL-6R Rabbit Recombinant Antibody [clone: DM152]
Antibodies
Recombinant rabbit monoclonal [DM152] antibody to IL-6R for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: IL-6R
Clonality: Recombinant
Clone: DM152
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR