Anti-Eph Receptor A3 Rabbit Recombinant Antibody [clone: DM162]

Supplier: ANTIBODIES.COM LLC
A318546-100 A318546-500
77186-822EA 1111.21 CAD
77186-822 77186-824
Anti-Eph Receptor A3 Rabbit Recombinant Antibody [clone: DM162]
Antibodies
Recombinant rabbit monoclonal [DM162] antibody to Eph Receptor A3 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: Eph Receptor A3
Clonality: Recombinant
Clone: DM162
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR