Anti-Carbonic Anhydrase 9/CA9 Rabbit Recombinant Antibody [clone: DM171]

Supplier: ANTIBODIES.COM LLC
A318538-100 A318538-500
77186-790EA 1111.21 CAD
77186-790 77186-792
Anti-Carbonic Anhydrase 9/CA9 Rabbit Recombinant Antibody [clone: DM171]
Antibodies
Recombinant rabbit monoclonal [DM171] antibody to Carbonic Anhydrase 9/CA9 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: Carbonic Anhydrase 9/CA9
Clonality: Recombinant
Clone: DM171
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR