Anti-MRP1 Mouse Monoclonal Antibody [clone: MRP1/1344]

Supplier: ANTIBODIES.COM LLC
A249379-100
76964-334EA 877.69 CAD
76964-334
Anti-MRP1 Mouse Monoclonal Antibody [clone: MRP1/1344]
Antibodies
Mouse monoclonal [MRP1/1344] antibody to MRP1 for Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone: MRP1/1344
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR