Anti-Carcinoembryonic Antigen Mouse Monoclonal Antibody [clone: COL-1 + CEA31 + C66/261]

Supplier: Antibodies.com LLC
A248136-100
76956-806EA 854.39 CAD
76956-806
Anti-Carcinoembryonic Antigen Mouse Monoclonal Antibody [clone: COL-1 + CEA31 + C66/261]
Antibodies
Mouse monoclonal [COL-1 + CEA31 + C66/261] antibody to Carcinoembryonic Antigen for Flow Cytometry, IF and IHC-P with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF, IHC-P
  • Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 0.5 to 1 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Carcinoembryonic Antigen
Clonality: Monoclonal
Clone: COL-1 + CEA31 + C66/261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR