Anti-UIS4 Goat Polyclonal Antibody

Supplier: Antibodies.com
A121573-1 A121573-400
76835-952EA 1489.78 CAD
76835-952 76834-378
Anti-UIS4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to UIS4 for IF with samples derived from Plasmodium berghei.

  • Validated Applications: IF
  • Recommended Dilutions: IF: 1:250 to 1:1000

Type: Primary
Antigen: UIS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Plasmodium berghei
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR