Anti-Galectin 3 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A104321-100 A104321-50
77062-866EA 877.69 CAD
77062-866 77067-978
Anti-Galectin 3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Galectin 3 for WB and ICC/IF with samples derived from Human, Rat, Mouse and Porcine.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:5000, ICC/IF: 1:2000

Type: Primary
Antigen: Galectin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse;Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR