Anti-Fibrillarin Mouse Monoclonal Antibody [clone: 4A4]

Supplier: ANTIBODIES.COM LLC
A104318-100 A104318-50
77066-106EA 877.69 CAD
77066-106 77065-610
Anti-Fibrillarin Mouse Monoclonal Antibody [clone: 4A4]
Antibodies
Mouse monoclonal (4A4) antibody to Fibrillarin for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse, Horse, Canine and Monkey.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:2000, ICC/IF: 1:1000 to 1:5000, IHC: 1:1000 to 1:5000

Type: Primary
Antigen: Fibrillarin
Clonality: Monoclonal
Clone: 4A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse;Horse;Dog;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR