Anti-GFAP Mouse Monoclonal Antibody [clone: ABT-GFAP]

Supplier: ANTIBODIES.COM LLC
A100021-100 A100021-50
77067-004EA 765.04 CAD
77067-004 77067-766
Anti-GFAP Mouse Monoclonal Antibody [clone: ABT-GFAP]
Antibodies
Mouse monoclonal (ABT-GFAP) antibody to GFAP for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: ABT-GFAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR