Anti-MCM2 Mouse Monoclonal Antibody [clone: ABT-MCM2]

Supplier: ANTIBODIES.COM LLC
A100019-50 A100019-100
77065-068EA 563.74 CAD
77065-068 77062-278
Anti-MCM2 Mouse Monoclonal Antibody [clone: ABT-MCM2]
Antibodies
Mouse monoclonal (ABT-MCM2) antibody to MCM2 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: MCM2
Clonality: Monoclonal
Clone: ABT-MCM2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR