Anti-Tartrate Resistant Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-TRAP]

Supplier: ANTIBODIES.COM LLC
A100013-50 A100013-100
77070-414EA 563.74 CAD
77070-414 77069-394
Anti-Tartrate Resistant Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-TRAP]
Antibodies
Mouse monoclonal (ABT-TRAP) antibody to TRAP for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Tartrate Resistant Acid Phosphatase
Clonality: Monoclonal
Clone: ABT-TRAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR