Anti-Hsp60 Mouse Monoclonal Antibody [clone: ABT-HSP60]

Supplier: ANTIBODIES.COM LLC
A99687-100 A99687-50
77071-716EA 765.04 CAD
77071-716 77080-694
Anti-Hsp60 Mouse Monoclonal Antibody [clone: ABT-HSP60]
Antibodies
Mouse monoclonal (ABT-HSP60) antibody to HSP60 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Hsp60
Clonality: Monoclonal
Clone: ABT-HSP60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR