Anti-TUG Rabbit Monoclonal Antibody [Clone: ARC2566]

Supplier: ANTIBODIES.COM LLC
A306667-100
77222-942EA 781.15 CAD
77222-942 77222-944
Anti-TUG Rabbit Monoclonal Antibody [Clone: ARC2566]
Antibodies
Rabbit monoclonal [ARC2566] antibody to TUG for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: TUG
Clonality: Monoclonal
Clone: ARC2566
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR