Anti-FABP7 Mouse Monoclonal Antibody [Clone: 2A84]

Supplier: Antibodies.com
A270549-100 A270549-50
77066-214EA 877.69 CAD
77066-214 77068-864
Anti-FABP7 Mouse Monoclonal Antibody [Clone: 2A84]
Antibodies
Mouse monoclonal [2A84] antibody to FABP7 for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1,000 to 1:2,000, ICC/IF: 1:1,000, IHC: 1:1,000

Type: Primary
Antigen: FABP7
Clonality: Monoclonal
Clone: 2A84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR